Assay Details
Gene Symbol: RNF135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023653703.1 | XP_023509471.1 | 961 | 60 | ||
GenBank mRNA | JL630659.1 | - | 2 - 3 | 813 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
