Assay Details
Gene Symbol: COPRS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023653050.1 | XP_023508818.1 | 238 | 61 | ||
XM_023653051.1 | XP_023508819.1 | 381 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
