Assay Details
Gene Symbol: ALDOC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504145.5 | XP_001504195.1 | 475 | 71 | ||
XM_005597616.3 | XP_005597673.1 | 514 | 71 | |||
XM_005597617.3 | XP_005597674.1 | 486 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
