Assay Details
Gene Symbol: PROCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003362427.4 | XP_003362475.1 | 525 | 69 | ||
XM_005597622.3 | XP_005597679.1 | 418 | 69 | |||
XM_014742221.2 | XP_014597707.1 | 601 | 69 | |||
XM_014742225.2 | XP_014597711.1 | 360 | 69 | |||
XM_023653107.1 | XP_023508875.1 | 336 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
