Assay Details
Gene Symbol: TRAF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023653113.1 | XP_023508881.1 | 621 | 56 | ||
XM_023653114.1 | XP_023508882.1 | 879 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
