Assay Details
Gene Symbol: FAM222B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014727579.2 | XP_014583065.2 | 126 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014727579.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
