Assay Details
Gene Symbol: CRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023653196.1 | XP_023508964.1 | 383 | 111 | ||
XM_023653197.1 | XP_023508965.1 | 385 | 111 | |||
XM_023653198.1 | XP_023508966.1 | 386 | 111 | |||
XM_023653199.1 | XP_023508967.1 | 382 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
