Assay Details
Gene Symbol: PAFAH1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023653240.1 | XP_023509008.1 | 1984 | 79 | ||
XM_023653241.1 | XP_023509009.1 | 1537 | 79 | |||
XM_023653242.1 | XP_023509010.1 | 1782 | 79 | |||
XM_023653243.1 | XP_023509011.1 | 1367 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
