Assay Details
Gene Symbol: NCBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504729.4 | XP_001504779.1 | 311 | 65 | ||
XM_005597667.3 | XP_005597724.1 | 311 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
