Assay Details
Gene Symbol: UBE2G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023653287.1 | XP_023509055.1 | 454 | 87 | ||
XM_023653288.1 | XP_023509056.1 | 457 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
