Assay Details
Gene Symbol: CXCL16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504756.6 | XP_001504806.1 | 802 | 73 | ||
XM_023653383.1 | XP_023509151.1 | 669 | 73 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CXCL16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504756.6 | XP_001504806.1 | 802 | 73 | ||
XM_023653383.1 | XP_023509151.1 | 669 | 73 |