Assay Details
Gene Symbol: PLSCR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503068.5 | XP_001503118.2 | 819 | 61 | ||
XM_005597741.3 | XP_005597798.1 | 815 | 61 | |||
XM_014728519.2 | XP_014584005.1 | 1031 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
