Assay Details
Gene Symbol: DHRS7C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503245.6 | XP_001503295.2 | 314 | 61 | ||
XM_005597780.3 | XP_005597837.1 | 314 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
