Assay Details
Gene Symbol: PEMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005597842.3 | XP_005597899.3 | 368 | 66 | ||
XM_005597844.3 | XP_005597901.1 | 376 | 66 | |||
XM_023653677.1 | XP_023509445.1 | 360 | 66 | |||
XM_023653678.1 | XP_023509446.1 | 444 | 66 | |||
XM_023653679.1 | XP_023509447.1 | 382 | 66 | |||
XM_023653680.1 | XP_023509448.1 | 320 | 66 | |||
XM_023653681.1 | XP_023509449.1 | 552 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
