Assay Details
Gene Symbol: PRR5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488316.4 | XP_001488366.1 | 869 | 64 | ||
XM_014729424.2 | XP_014584910.1 | 748 | 64 | |||
XM_023653887.1 | XP_023509655.1 | 2821 | 64 | |||
XM_023653888.1 | XP_023509656.1 | 741 | 64 | |||
XM_023653889.1 | XP_023509657.1 | 694 | 64 | |||
XM_023653890.1 | XP_023509658.1 | 1423 | 64 | |||
XM_023653891.1 | XP_023509659.1 | 2836 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
