Assay Details
Gene Symbol: MDK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023653975.1 | XP_023509743.1 | 368 | 62 | ||
XM_023653976.1 | XP_023509744.1 | 263 | 62 | |||
XM_023653977.1 | XP_023509745.1 | 263 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
