Assay Details
Gene Symbol: SPI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491380.5 | XP_001491430.1 | 689 | 67 | ||
XM_005598097.3 | XP_005598154.1 | 692 | 67 | |||
GenBank mRNA | JN979559.1 | - | 4 - 5 | 591 | 67 | |
JN979560.1 | - | 4 - 5 | 588 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
