Assay Details
Gene Symbol: UBE2L6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498309.5 | XP_001498359.1 | 292 | 91 | ||
XM_005598144.3 | XP_005598201.1 | 618 | 91 | |||
XM_005598145.3 | XP_005598202.1 | 347 | 91 | |||
XM_005598146.3 | XP_005598203.1 | 398 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
