Assay Details
Gene Symbol: LOC100068374 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498226.3 | XP_001498276.2 | 127 | 55 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001498226.3 transcripts.
Set Membership: |
![]() ![]() |
