Assay Details
Gene Symbol: LOC100066603 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504695.3 | XP_001504745.1 | 175 | 71 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001504695.3 transcripts.
Set Membership: |
![]() ![]() |
