Assay Details
Gene Symbol: DTX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001915461.4 | XP_001915496.2 | 2163 | 65 | ||
XM_023654102.1 | XP_023509870.1 | 1752 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
