Assay Details
Gene Symbol: MS4A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493378.5 | XP_001493428.2 | 775 | 74 | ||
XM_005598200.3 | XP_005598257.1 | 827 | 74 | |||
XM_023654124.1 | XP_023509892.1 | 692 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
