Assay Details
Gene Symbol: FADS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023654187.1 | XP_023509955.1 | 1477 | 55 | ||
XM_023654188.1 | XP_023509956.1 | 1386 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
