Assay Details
Gene Symbol: PLAAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005598258.3 | XP_005598315.2 | 452 | 116 | ||
XM_005598259.3 | XP_005598316.2 | 451 | 116 |

More Information
Set Membership: |
![]() ![]() |
