Assay Details
Gene Symbol: KYAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495159.6 | XP_001495209.2 | 381 | 67 | ||
XM_023641709.1 | XP_023497477.1 | 411 | 67 | |||
XM_023641710.1 | XP_023497478.1 | 368 | 67 | |||
XM_023641712.1 | XP_023497480.1 | 433 | 67 | |||
XM_023641713.1 | XP_023497481.1 | 294 | 67 | |||
GenBank mRNA | JL617536.1 | - | 4 - 5 | 346 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
