Assay Details
Gene Symbol: BCL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495178.5 | XP_001495228.2 | 338 | 99 | ||
GenBank mRNA | JL616141.1 | - | 1 - 2 | 105 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
