Assay Details
Gene Symbol: SYDE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003365059.4 | XP_003365107.2 | 1037 | 73 | ||
XM_023641743.1 | XP_023497511.1 | 1040 | 73 | |||
XM_023641744.1 | XP_023497512.1 | 1041 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
