Assay Details
Gene Symbol: LEPROT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023641938.1 | XP_023497706.1 | 180 | 117 | ||
XM_023641939.1 | XP_023497707.1 | 180 | 117 | |||
XM_023641940.1 | XP_023497708.1 | 180 | 117 | |||
XM_023641941.1 | XP_023497709.1 | 122 | 117 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023641941.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
