Assay Details
Gene Symbol: LEPROT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023641939.1 | XP_023497707.1 | 363 | 94 | ||
XM_023641941.1 | XP_023497709.1 | 305 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
