Assay Details
Gene Symbol: UBE2U | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499644.6 | XP_001499694.4 | 899 | 95 | ||
XM_023641955.1 | XP_023497723.1 | 901 | 95 | |||
XM_023641956.1 | XP_023497724.1 | 908 | 95 | |||
XM_023641957.1 | XP_023497725.1 | 906 | 95 | |||
XM_023641959.1 | XP_023497727.1 | 906 | 95 | |||
XM_023641960.1 | XP_023497728.1 | 904 | 95 | |||
XM_023641961.1 | XP_023497729.1 | 904 | 95 | |||
XM_023641962.1 | XP_023497730.1 | 904 | 95 | |||
XM_023641963.1 | XP_023497731.1 | 901 | 95 | |||
XM_023641964.1 | XP_023497732.1 | 902 | 95 | |||
XM_023641965.1 | XP_023497733.1 | 900 | 95 | |||
XM_023641967.1 | XP_023497735.1 | 299 | 95 | |||
XM_023641969.1 | XP_023497737.1 | 267 | 95 | |||
XM_023641970.1 | XP_023497738.1 | 397 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
