Assay Details
Gene Symbol: ITGB3BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014740089.2 | XP_014595575.2 | 461 | 78 | ||
XM_023641986.1 | XP_023497754.1 | 525 | 78 | |||
XM_023641987.1 | XP_023497755.1 | 505 | 78 | |||
XM_023641988.1 | XP_023497756.1 | 589 | 78 | |||
XM_023641989.1 | XP_023497757.1 | 519 | 78 | |||
XM_023641990.1 | XP_023497758.1 | 385 | 78 | |||
XM_023641991.1 | XP_023497759.1 | 357 | 78 | |||
XM_023641992.1 | XP_023497760.1 | 956 | 78 | |||
XM_023641993.1 | XP_023497761.1 | 294 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
