Assay Details
Gene Symbol: VEGFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023654288.1 | XP_023510056.1 | 731 | 66 | ||
XM_023654290.1 | XP_023510058.1 | 718 | 66 | |||
XM_023654291.1 | XP_023510059.1 | 632 | 66 | |||
XM_023654292.1 | XP_023510060.1 | 618 | 66 | |||
XM_023654293.1 | XP_023510061.1 | 715 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
