Assay Details
Gene Symbol: FKBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489557.5 | XP_001489607.2 | 371 | 57 | ||
XM_003362625.4 | XP_003362673.2 | 324 | 57 | |||
XM_023654295.1 | XP_023510063.1 | 380 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
