Assay Details
Gene Symbol: GPR137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005598273.3 | XP_005598330.1 | 834 | 89 | ||
XM_005598274.3 | XP_005598331.1 | 951 | 89 | |||
XM_005598275.2 | XP_005598332.1 | 951 | 89 | |||
XM_023654300.1 | XP_023510068.1 | 557 | 89 | |||
XM_023654301.1 | XP_023510069.1 | 951 | 89 | |||
XM_023654302.1 | XP_023510070.1 | 951 | 89 | |||
XM_023654303.1 | XP_023510071.1 | 951 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
