Assay Details
Gene Symbol: ESRRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023654305.1 | XP_023510073.1 | 880 | 76 | ||
XM_023654307.1 | XP_023510075.1 | 640 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
