Assay Details
Gene Symbol: CFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494534.6 | XP_001494584.1 | 152 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001494534.6 transcripts.
Set Membership: |
![]() ![]() |
