Assay Details
Gene Symbol: ALDH3B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005598349.3 | XP_005598406.2 | 1344 | 94 | ||
XM_005598350.3 | XP_005598407.1 | 1345 | 94 | |||
XM_014729726.2 | XP_014585212.1 | 1108 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
