Assay Details
Gene Symbol: CRACR2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005598396.3 | XP_005598453.1 | 933 | 84 | ||
XM_005598397.3 | XP_005598454.1 | 936 | 84 | |||
XM_005598398.3 | XP_005598455.1 | 934 | 84 | |||
XM_014729766.2 | XP_014585252.1 | 936 | 84 | |||
XM_023654783.1 | XP_023510551.1 | 868 | 84 | |||
XM_023654784.1 | XP_023510552.1 | 866 | 84 | |||
XM_023654785.1 | XP_023510553.1 | 961 | 84 | |||
XM_023654786.1 | XP_023510554.1 | 1542 | 84 | |||
XM_023654787.1 | XP_023510555.1 | 1449 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
