Assay Details
Gene Symbol: MOB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488276.6 | XP_001488326.3 | 588 | 55 | ||
XM_023654825.1 | XP_023510593.1 | 389 | 55 | |||
XM_023654826.1 | XP_023510594.1 | 597 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() |
