Assay Details
Gene Symbol: DUSP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023654827.1 | XP_023510595.1 | 6689 | 74 | ||
XM_023654828.1 | XP_023510596.1 | 996 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
