Assay Details
Gene Symbol: PGGHG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023654863.1 | XP_023510631.1 | 1760 | 63 | ||
XM_023654864.1 | XP_023510632.1 | 1683 | 63 | |||
XM_023654865.1 | XP_023510633.1 | 1629 | 63 | |||
XM_023654866.1 | XP_023510634.1 | 1553 | 63 | |||
XM_023654867.1 | XP_023510635.1 | 1763 | 63 | |||
XM_023654868.1 | XP_023510636.1 | 1432 | 63 | |||
XM_023654869.1 | XP_023510637.1 | 1764 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
