Assay Details
Gene Symbol: BUD31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494750.6 | XP_001494800.1 | 371 | 96 | ||
XM_023655308.1 | XP_023511076.1 | 404 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
