Assay Details
Gene Symbol: ARPC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023655301.1 | XP_023511069.1 | 822 | 60 | ||
XM_023655302.1 | XP_023511070.1 | 666 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
