Assay Details
Gene Symbol: AIMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005598523.3 | XP_005598580.1 | 594 | 74 | ||
XM_014729804.2 | XP_014585290.1 | 371 | 74 | |||
XM_023655258.1 | XP_023511026.1 | 481 | 74 | |||
XM_023655259.1 | XP_023511027.1 | 344 | 74 | |||
XM_023655260.1 | XP_023511028.1 | 359 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
