Assay Details
Gene Symbol: NUDT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001915099.4 | XP_001915134.1 | 220 | 76 | ||
XM_023655148.1 | XP_023510916.1 | 228 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
