Assay Details
Gene Symbol: GET4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023655072.1 | XP_023510840.1 | 939 | 57 | ||
XM_023655073.1 | XP_023510841.1 | 938 | 57 | |||
XM_023655075.1 | XP_023510843.1 | 791 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
