Assay Details
Gene Symbol: RCC1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023616809.1 | XP_023472577.1 | 1403 | 79 | ||
XM_023616810.1 | XP_023472578.1 | 1244 | 79 | |||
XM_023616811.1 | XP_023472579.1 | 1229 | 79 | |||
GenBank mRNA | JL628026.1 | - | 1 - 2 | 91 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
