Assay Details
Gene Symbol: ACHE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005598649.3 | XP_005598706.1 | 2316 | 95 | ||
XM_014729888.2 | XP_014585374.2 | 1543 | 95 | |||
XM_023655373.1 | XP_023511141.1 | 1543 | 95 | |||
XM_023655374.1 | XP_023511142.1 | 1211 | 95 | |||
XM_023655375.1 | XP_023511143.1 | 1214 | 95 | |||
XM_023655376.1 | XP_023511144.1 | 2319 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
