Assay Details
Gene Symbol: BCL7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014729899.2 | XP_014585385.2 | 518 | 70 | ||
XM_023655440.1 | XP_023511208.1 | 349 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
