Assay Details
Gene Symbol: SBDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499997.6 | XP_001500047.1 | 851 | 85 | ||
GenBank mRNA | JL640398.1 | - | 1 - 2 | 160 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
